missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ NanoDrop™ Lite Plus Printer Accessory

Use an optional printer with your NanoDrop™ Lite Plus Spectrophotometer. Print data on freezer-compatible labels.

Supplier:  Thermo Scientific™ NDLPPRNTRMOD

Catalog No. NDLPPRNTRM


Only null left

Description

Description

Use an optional printer with your NanoDrop™ Lite Plus Spectrophotometer. Print data on freezer-compatible labels.

Print sample data on a cryogenic label

  • Sample number
  • Date
  • Time of Measurement
  • Sample Name
  • Assay
  • Concentration
  • Purity Ratio

For use with the NanoDrop Lite Spectrophotometer

Specifications

Specifications

Printer Accessory for NanoDrop Lite Plus
NanoDrop Lite Plus Spectrophotometer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions