Learn More
Clontech Laboratories PrimeSTAR® GXL DNA Polymerase, 250 units
Supplier: Clontech Laboratories R050A
A hot-start, high-fidelity PCR enzyme, PrimeSTAR GXL DNA Polymerase excels in reactions with GC-rich templates, excess template, and long amplicons up to 30 kb (GXL). The polymerase is supplied with separate tubes of optimized buffer (Mg2+ plus), and dNTPs. 250 units