missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Primary Rat Hippocampus Neurons

Catalog No. A1084101
Encompass
Change view
Click to view available options
Quantity:
1 x 106 cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1084101 1 x 106 cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1084101 Supplier Gibco™ Supplier No. A1084101

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Primary Rat Hippocampus Neurons

GIBCO Primary Rat Hippocampus Neurons are isolated from day-18 rat embryos and cryopreserved in liquid nitrogen. Each vial contains 1x10e6 cells⁄ml viable cells (post-thaw). With the cryopreserved Primary Rat Hippocampus Neurons (E-18), Invitrogen offers neuroscience researchers a flexible, ready-to-use, and quality alternative to acutely isolated neurons. Besides being convenience, these products are developed through vigorous internal research to afford comparable properties, such as viability and cell function, with cells that are acutely isolated. They are also highly pure cells containing minimum amount of astrocytes and other glial cells, and are with minimal lot-to-lot variability because of large single lot manufacturing. Unlike the other commercially available primary rat hippocampus neurons that generally yield about 30% or lower post-thaw viability, Invitrogen cells consistently give post-thaw viability in the range of 50-80%.

Specifications

Content And Storage Store in liquid nitrogen, vapor-phase.
Age Fetal
Cell Type Neural Cells
Culture Type Mammalian Cell Culture
Form Cryopreserved
Species Rat
Tissue Hippocampus
Donor Attributes Normal
No. of Cells 1 x 106 cells
Product Type Rat Hippocampus Neuron
Quantity 1 x 106 cells
Tested For Sterility, Viability, Mycoplasma
Show More Show Less
I'm currently culturing neurons. Which media system do you recommend using in conjunction with the Poly-D-Lysine matrix?

We recommend using the B-27 Plus Neuronal Culture System when culturing neurons on Poly-D-Lysine. The B-27 Plus Neuronal Culture System is a complete media system that includes the B-27 Plus Supplement and the Neurobasal Plus Medium.

What cell types will Poly-D-Lysine support as an attachment matrix?

The Poly-D-Lysine matrix has successfully supported the attachment of primary rodent neurons, iPSC-derived neurons and PC12 cells.

Can primary rat neurons be cryopreserved again after they are cultured?

Primary rat neurons cannot be cryopreserved again because they will not revive.

Can primary rat neurons proliferate?

No, primary rat neurons do not proliferate.

Why does the manual for primary rat neurons have the instruction "Do not expose neurons to air at any time"?

Neurons tend to lift off from the complete medium change, which exposes cells to air. By changing half of the medium, the disturbance can be minimized.

For Research Use Only. Not for use in diagnostic procedures.