missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Primair™ Headgear Replacement for PA701HED PAPR

Catalog No. 19085583
Change view
Click to view available options
Replacement
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19085583 Replacement
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19085583 Supplier Honeywell™ Supplier No. PA103

Replacement part for use with the PA701HED PAPR system.

  • Replacement part
  • Headgear
  • Black
  • For use with PA701HED

Specifications

Color Black
Disposable Reusable
Product Type Headgear
Size One Size
For Use With (Equipment) Primair™ PA701HED Series PAPR
Model PA103