missing translation for 'onlineSavingsMsg'
Learn More

Bio-Serv™ Prima-Treats™ Animal Diet

Catalog No. 14726698
Encompass
Change view
Click to view available options
Flavor:
Apple
Banana
Blueberry
Orange
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flavor
14726698 Blueberry
14726454 Apple
14726456 Banana
14726460 Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14726698 Supplier Bio-Serv™ Supplier No. F0293

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Healthy treat used for training or positive reward following a less desirable procedure.

  • Comes in variety of flavors
  • Nutritionally complete treat that compliments enrichment programs
  • Less labor intensive and more sanitary than perishable fruit
  • Excellent source of vitamin C
  • Nutritionally assayed
  • Weight: 5g (0.18 oz.)
  • Shape: Wafer
  • Packaging: 250/Jar and 1000/Box.

Specifications

Product Type Animal Diet
Calories 16.4 kcal/Wafer
Components Dextrose, Casein, Sucrose, Cellulose, Mineral Mix, Canola Oil, Natural and Artificial Flavors, Magnesium Silicate, Choline Dihydrogen Citrate, Vitamin Mix, Calcium Silicate, Ascorbic Acid, DL-Methionine, Magnesium Stearate, Food Dye, BHA
Flavor Blueberry
Particle Shape Wafer
Quantity 1000/Box
Storage Requirements Cool dry storage
Diameter (English) 1 in.
Diameter (Metric) 2.54 cm