Learn More
Abcam Inc Prestained Protein Ladder - E x tra broad molecular weight (5 - 245 kDa), 500UL
Catalog No. 502584840
Encompass
5-245 kDa prestained protein ladder is a three-color protein standard with 13 prestained proteins. - Suitable for western blot (WB), SDS-PAGE - Designed for monitoring protein separation - Supplied in gel loading buffer and is ready to use
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 502584840 Supplier Abcam Inc Supplier No. AB116029500UL
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
5-245 kDa prestained protein ladder is a three-color protein standard with 13 prestained proteins. - Suitable for western blot (WB), SDS-PAGE - Designed for monitoring protein separation - Supplied in gel loading buffer and is ready to use
The product is subject to the following: Abcam Restricted Use Statement