Learn More
Abcam Inc Prestained Protein Ladder - E x tra broad molecular weight (3 - 260 kDa), 500UL
Catalog No. 502699683
Encompass
Prestained Protein Ladder AB313996 is a combination of 11 pre-stained proteins with molecular weights from 3 to 260 kDa. This prestained protein ladder keeps track of the size and separation of proteins during SDS-polyacrylamide gel electrophoresis, approximating the target protein size and validating the Western transfer efficiency on PVDF, nylon, or nitrocellulose membranes.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 502699683 Supplier Abcam Inc Supplier No. AB313996500UL
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Prestained Protein Ladder AB313996 is a combination of 11 pre-stained proteins with molecular weights from 3 to 260 kDa. This prestained protein ladder keeps track of the size and separation of proteins during SDS-polyacrylamide gel electrophoresis, approximating the target protein size and validating the Western transfer efficiency on PVDF, nylon, or nitrocellulose membranes.
The product is subject to the following: Abcam Restricted Use Statement