missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ PrepSEQ™ Elution Buffer

Catalog No. 4400784C
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4400784C 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4400784C Supplier Applied Biosystems™ Supplier No. 4400784C*
Get an Extra Analytical Consumable with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PrepSEQ Elution Buffer, a component of the PrepSEQ Nucleic Acid Extraction Kit, is offered separately here for those kit users who need additional elution buffer.

PrepSEQ Elution Buffer, a component of the PrepSEQ Nucleic Acid Extraction Kit (Cat. No. 4413686), is offered separately here for those kit users who need additional elution buffer. As with other products in the Pharma Analytics Sample Preparation product line, PrepSEQ Elution Buffer:

  • Is completely supported by our global manufacturing and distribution centers
  • Has a Certificate of Analysis available
  • Has a shelf-life of up to 12 months

Specifications

Chemical Name or Material Elution Buffer
Physical Form Liquid
Quantity 25 mL

For Research Use Only. Not for use in diagnostic procedures.