missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ PrepSEQ™ Elution Buffer

Catalog No. 361054959
Encompass
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
361054959 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361054959 Supplier Applied Biosystems™ Supplier No. 4400784C

missing translation for 'validMainframeMsg'

PrepSEQ Elution Buffer, a component of the PrepSEQ Nucleic Acid Extraction Kit, is offered separately here for those kit users who need additional elution buffer.

PrepSEQ Elution Buffer, a component of the PrepSEQ Nucleic Acid Extraction Kit (Cat. No. 4413686), is offered separately here for those kit users who need additional elution buffer. As with other products in the Pharma Analytics Sample Preparation product line, PrepSEQ Elution Buffer:

  • Is completely supported by our global manufacturing and distribution centers
  • Has a Certificate of Analysis available
  • Has a shelf-life of up to 12 months

Specifications

Quantity 25 mL
Content And Storage Store at room temperature.
Form Liquid
Product Type Elution Buffer

For Research Use Only. Not for use in diagnostic procedures.