Learn More
MilliporeSigma™ PrePak L2 Pretreatment Pack
Catalog No. PRPK0L002
Change view
Click to view available options
Type:
Water Purification Consumables
Catalog No. Type
PRPK0L002 Water Purification Consumables
1 options
Catalog No. PRPK0L002 Supplier MilliporeSigma™ Supplier No. PRPK0L002
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Lab Water purification system portfolio offers a broad range of pure and ultrapure water purification systems and services.
  • Designed for scientists working in pharmaceutical, clinical, academic, industrial, research, and government laboratories
  • Can be used in both validated and non-validated environments
  • The PrePack has been designed to pretreat water by removing large particles from tap water
  • Benefits
    • Optimal protection of the water system
    • Ensures system lifetime and performance are optimized
Specifications
Type Water Purification Consumables
For Use With (Equipment) Elix™ 20/35/70/100 Systems and RiOs™ 30/50/100/150/200