Learn More
Eisco™ Premium Two Prong Clamp
Catalog No. S13909
Change view
Click to view available options
Color:
Blue
Catalog No. Color
S13909 Blue
1 options
Catalog No. S13909 Supplier Eisco™ Supplier No. PRCMP05
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Choose durable lab equipment that will last for years. The Premium Two Prong Clamp is an industrial two prong clamp for burettes, flasks, and beaker clamps.
  • Clamp constructed of powder coated zinc alloy, 2 prongs attached to aluminum rod
  • Maximum clamp opening is 12 cm
  • 14 cm connecting rod
  • Completely metal constructions, thread fitted actuating joints
  • 10 mm diameter connecting rod
Specifications
Autoclavable Autoclavable
Type 2-Prong Clamp
Material Nickel-plated
Color Blue
Jaw Type 2-Prong
Coating Vinyl