missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eisco™ Premium Two Prong Clamp w/ Boss Head Attachment

Choose durable lab equipment that will last for years. The EISCO Premium Two Prong Clamp w/ Boss Head Attachment is an industrial,rubber-coated clamp for burettes, flasks, and beaker clamps with a swivel boss head attachment.

Supplier:  Eisco™ PRCMP05BHEMD

Catalog No. S13910



Description

Description

  • Maximum clamp opening is 10.5 cm
  • Swivel between clamp and boss head
  • Boss accommodates to 19mm rod
Specifications

Specifications

Autoclavable
Blue
SDS
Documents

Documents

Promotions

Promotions