Learn More
Eisco™ Premium Three Finger Clamp w/ Dual Screw Adjustments
Choose durable lab equipment that will last for years. The EISCO Premium Three Finger Clamp w/ Dual Screw Adjustments is an industrial three finger, rubber-coated extension lab clamp with dual screw adjustments.
Supplier:  Eisco™ EISPRCMP06
Catalog No. S13911
Only null left
Explore available promotions
Description
Description
  • Heavy duty spring loaded clamp jaw with dual screw adjustment to fine tune any fitting
  • Made of steel and powder coated alloy with all metal adjustment screws
  • 3 rubber coated clamp fingers
  • 6cm maximum opening clamp
  • 12.5cm steel connecting rod
Specifications
Specifications
Clamp
6 cm max. opening
3-Prong
Steel, Powder-Coated Alloy, Metal (Screws)
Blue
Product Suggestions
Product Suggestions