missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ Premium Split Cowhide Leather Palm Gloves with Aramid Fiber Stitching

Catalog No. p-7214578
Change view
Click to view available options
Size:
Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
1146260B Large
1146260C X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 1146260B Supplier Wells Lamont™ Supplier No. Y3015L
Only null left

Premium Split Cowhide Leather Palm Gloves with Aramid Fiber Stitching are constructed from superior quality grade leather, and stitched with aramid fibers for increased strength and durability.

  • Sewn with aramid fiber thread for increased strength at the seams
  • Premium quality split cowhide extends the glove's usage cycle
  • An extended gauntlet cuff provides protection up to the forearm
  • Leather knuckle strap
  • Canvas back material
  • Wing thumb

Specifications

Back Material Canvas
Color Natural, Gray
Grade Superior
Hand Specific
Material Shoulder Split Cowhide Leather, Aramid Fiber
Product Type Leather Palm Gloves
Size Large
Glove Cuff Color Natural
Glove Cuff Style Gauntlet
Hem Color Brown
Series Y3015
Thumb Style Wing
Show More Show Less