missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ Premium Quality Split Cowhide Leather Palm Gloves with Aramid Fiber Stitching

Premium Quality Split Cowhide Leather Palm Gloves are superior quality grade leather gloves sewn with aramid fibers to increase durability and extend the glove's useful life. A leather knuckle strap protects the back of the hand, while the canvas back increases air circulation.

$184.63 - $184.63

Specifications

Back Material Canvas
Color Natural, Gray
Grade Premium
Hand Specific
Material Shoulder Split Cowhide Leather, Aramid Fiber
View More Specs

Products 1
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19823911
View Documents
Wells Lamont™
Y3014XL
X-Large
Dozen of 12 Pair for $184.63
Only null left
 
Description

Description

  • Sewn with Aramid Fiber thread for increased strength at the seams
  • Premium quality split cowhide extends the glove's usage cycle
  • Leather knuckle strap
  • Canvas back material
  • Safety cuff
  • Wing thumb
Specifications

Specifications

Canvas
Premium
Shoulder Split Cowhide Leather, Aramid Fiber
Natural
Y3014
Natural, Gray
Specific
Leather Palm Gloves
Safety
Wing
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'