Learn More
Eisco™ Premium Practice Suture Kit
Catalog No. p-7201186
Change view
Click to view available options
Product Type:
Complete Practice Kit
Catalog No. Product Type
S27477 Complete Practice Kit
1 options
Catalog No. S27477 Supplier Eisco™ Supplier No. BI0201903
Only null left
Designed by a physician, the Practice Suture Kit includes everything needed to practice suturing skills, including a variety of wound types and needle sizes. The innovative plastic suture pad is durable and can be reused many times.
  • Designed for practicing suturing skills
  • Endorsed by professionals for medical, nursing & veterinary students, as well as for surgical, emergency & anesthesia residents
  • Innovative Suture Pad is durable and does not tear, allowing each cut to be sutured multiple times
  • Features a realistic texture and a variety of common cuts and lacerations, irregular wounds, and Y-shaped wound for corner stitches
  • Eisco Exlusive: Cuts with irregular widths to approximate a wound that had been cleaned of debris, dead skin, and slits to practice securing chest tubes, catheters, drains, and central lines
  • Suture Pad Dimensions: (6.5 x 5.75 x 0.5 in.)
  • For practice and educational use only
Specifications
Product Type Complete Practice Kit
For Use With (Application) Suturing Skills
Includes Suture Pad with Wounds (1), Needle Holder (1), Toothed Forceps (1), Fine Suturing Scissors (1), Braided Silk Suture Varieties (0/0, 2/0, 3/0, 4/0, 5/0), 19 mm Half Circle Needles (4), 37 mm Half Circle Needle (1), Tubing Pieces (2)