Learn More
Gilson™ Premium Pipette Service for Multichannel Pipettes
Catalog No. SVC101200G
Change view
Click to view available options
Product Type:
Pipette Premium Lifetime Service
For Use With (Equipment):
Multi-channel Pipette
Service Offering:
After Repair Calibration
Calibration Points:
2 x 2 Measurements
Catalog No. Product Type For Use With (Equipment) Service Offering Calibration Points
SVC101200G Pipette Premium Lifetime Service Multi-channel Pipette After Repair Calibration 2 x 2 Measurements
1 options
Catalog No. SVC101200G Supplier Gilson™ Supplier No. SVC101200
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Gilson Premium Pipette Service is the most comprehensive level of service, providing users with reliable performance and predictable pricing. At least once per year Gilson comes with a lifetime warranty on the pipette and all necessary repairs are included at no additional cost.
  • Visual and Functional Inspection
  • Internal and External Cleaning
  • Leak Test
  • Gravimetric Check
  • Preventative Maintenance
  • Re-grease as needed
  • Pipette Label
  • Mail-In or Onsite Service Available
  • Calibration and Detailed Certificate
  • ISO 17025 Calibration Available
  • Lifetime Warranty
  • Free Repairs
  • 3 day turn around time
Specifications
For Use With (Equipment) Multi-channel Pipette
Certifications/Compliance ISO 17025 Accredited Service
Description Data collected after repair
Calibration Points 2 x 2 Measurements
Duration 3 Days
Product Type Pipette Premium Lifetime Service
Service Offering After Repair Calibration