missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Premium Foam-Insulated Leather Drivers Gloves

Catalog No. 19013379
Change view
Click to view available options
Size:
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19013379 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19013379 Supplier Fisherbrand™ Supplier No. 77208/L
Only null left

Constructed from top grain cowhide leather with foam insulation to keep hands warm during cold applications.

  • Ideal for cold weather applications such as farming, construction, and refrigeration
  • Straight thumb and shirred elastic back
  • Red foam lining adds comfort and warmth
  • Easy size identification by color and cuff
  • Gunn cut

Specifications

Color Natural
Grade Premium
Hand Specific
Material Top Grain Cowhide Leather
Product Type Driver Gloves
Size Large
Style Gunn
For Use With (Application) Farming, Construction, Machine Operation, and Refrigeration
Glove Cuff Style Open
Hem Color Brown
Liner Material Foam
Series 77-208
Length (English) 10.2 in.
Length (Metric) 26 cm
Show More Show Less