missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Premium Die Casted Boss Head Clamp

Catalog No. S13919
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S13919 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S13919 Supplier Eisco™ Supplier No. PRBH01

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ideal for connecting 16mm diameter rods and apparatus in tight places without sacrificing strength.

  • Threaded metal one piece clamping screws
  • All metal constructions to carry heavy loads and prevent torque damage
  • Able to hold rods up to 16mm in diameter
  • Holds rods orthogonal (90 degrees) to one another
  • Made from high grade zinc alloy with non toxic powder coating finish
  • With two stainless steel thumb screws

Specifications

Autoclavable Autoclavable
Type Adjustable Clamp
Material Zinc Die Cast, Powder Coated
Holds Holds rods up to 16 mm diameter
For Use With (Application) Holds rods orthogonal (90 degrees) to one another
Color Blue
Jaw Type Single Adjustment