missing translation for 'onlineSavingsMsg'
Learn More

Pregnenolone, 98%, Spectrum™ Chemical

$47898.15 - $119988.74

Chemical Identifiers

CAS 145-13-1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18612947
View Documents
Spectrum Chemical Mfg Cor
P14225KGBL
5 kg Poly Pail N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
18612945
View Documents
Spectrum Chemical Mfg Cor
P142225KGBL
25 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

145-13-1

Specifications

145-13-1
0.005
Poly Pail
+27° to +30°
Reagent
1
183° to 194°C
5 kg
0.98
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For use by qualified personnel for research and development or other authorized use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.