Learn More
Chemplex Industries PRECUT CIRCLES MYLAR 0.24MIL DFS Item
Supplier:  Chemplex Industries CMP256
CHEMPLEX THIN-FILM, PRECUT CIRCLES, Mylar(TM), 6.0u (0.24mil), 2.5 Dia. (6.4cm)
Catalog No. 50249336
Only null left
Explore available promotions