missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Precoated TLC and PLC Glass Plates

Catalog No. m1057460001
Change view
Click to view available options
Dimensions (L x W):
2.5 x 7.5 cm
5 x 10 cm
5 x 20 cm
10 x 20 cm
20 x 10 cm
20 x 20 cm
Quantity:
100/Pk.
12/Pk.
20/Pk.
25/Pk.
50/Pk.
Material:
Cellulose
Cellulose F
Kieselguhr F254
PEI Cellulose F
Silica 60
Silica 60 (0.5 mm)
Silica 60 (2 mm)
Silica Gel 60 F254
Silica Gel 60 F254 AMD, extra thin
Silica Gel 60 RP-18 F254 s
Silica Gel 60 RP-2 (silanized)
Silica Gel 60 RP-2 F254 (silanized)
Silica Gel 60 RP-8 F254
Silica Gel 60 RP-8 F254 s
Silica Gel 60 W F254 s
Type:
Cellulose TLC Plate
Classical Silica TLC Plate
Concentrating Zone (2.5 x 10 cm) TLC Plate
Concentrating Zone (2.5 x 20 cm) TLC Plate
Kieselguhr TLC Plate
RP-modified Silica TLC Plate
Silica Gel PLC Plate
Silica HPTLC Plate
Unmodified Silica Gel TLC Plate
28 product options available for selection
Product selection table with 28 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Quantity Material Type
M1057460001 20 x 20 cm 25/Pk. Silica Gel 60 RP-2 (silanized) RP-modified Silica TLC Plate
M1057210001 20 x 20 cm 25/Pk. Silica 60 Unmodified Silica Gel TLC Plate
M1153890001 20 x 20 cm 25/Pk. Silica Gel 60 RP-18 F254 s RP-modified Silica TLC Plate
M1117980001 20 x 20 cm 25/Pk. Silica Gel 60 F254 Concentrating Zone (2.5 x 20 cm) TLC Plate
M1153270001 2.5 x 7.5 cm 100/Pk. Silica Gel 60 F254 Unmodified Silica Gel TLC Plate
M1156830001 5 x 20 cm 50/Pk. Silica Gel 60 RP-18 F254 s RP-modified Silica TLC Plate
M1118460001 10 x 20 cm 50/Pk. Silica Gel 60 F254 Concentrating Zone (2.5 x 10 cm) TLC Plate
M1057450001 20 x 20 cm 12/Pk. Silica 60 (2 mm) Silica Gel PLC Plate
M1117640001 20 x 10 cm 25/Pk. Silica Gel 60 F254 AMD, extra thin Silica HPTLC Plate
M1164850001 20 x 20 cm 25/Pk. Silica Gel 60 W F254 s Classical Silica TLC Plate
M1057250001 20 x 20 cm 25/Pk. PEI Cellulose F Cellulose TLC Plate
M1057180001 20 x 20 cm 25/Pk. Cellulose F Cellulose TLC Plate
M1154240001 10 x 20 cm 50/Pk. Silica Gel 60 RP-8 F254 s RP-modified Silica TLC Plate
M1153260001 2.5 x 7.5 cm 100/Pk. Silica 60 Unmodified Silica Gel TLC Plate
M1057380001 20 x 20 cm 25/Pk. Kieselguhr F254 Kieselguhr TLC Plate
M1154230001 10 x 20 cm 50/Pk. Silica Gel 60 RP-18 F254 s RP-modified Silica TLC Plate
M1156840001 5 x 10 cm 25/Pk. Silica Gel 60 RP-8 F254 s RP-modified Silica TLC Plate
M1138940001 20 x 20 cm 20/Pk. Silica 60 (0.5 mm) Silica Gel PLC Plate
M1057280001 10 x 20 cm 50/Pk. Cellulose F Cellulose TLC Plate
M1057240001 5 x 20 cm 100/Pk. Silica 60 Unmodified Silica Gel TLC Plate
M1118450001 20 x 20 cm 25/Pk. Silica 60 Concentrating Zone (2.5 x 20 cm) TLC Plate
M1057470001 20 x 20 cm 25/Pk. Silica Gel 60 RP-2 F254 (silanized) RP-modified Silica TLC Plate
M1153880001 20 x 20 cm 25/Pk. Silica Gel 60 RP-8 F254 RP-modified Silica TLC Plate
M1057160001 20 x 20 cm 25/Pk. Cellulose Cellulose TLC Plate
M1118440001 10 x 20 cm 50/Pk. Silica 60 Concentrating Zone (2.5 x 10 cm) TLC Plate
M1056260001 10 x 20 cm 50/Pk. Silica 60 Unmodified Silica Gel TLC Plate
M1057300001 10 x 20 cm 50/Pk. Cellulose Cellulose TLC Plate
M1156850001 20 x 20 cm 25/Pk. Silica Gel 60 RP-8 F254 s RP-modified Silica TLC Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 28 options available.
28 options
Catalog No. M1057460001 Supplier MilliporeSigma™ Supplier No. 1.05746.0001

Average particle size is 15μm with a 5-20μm distribution

  • Choose from several media to match your separation requirements: silica gel 60 (with and without F254 fluorescent indicator), chemically modified silica gel (reversed phases RP-2, RP-8 and RP-18), aluminum oxide 60 with F254 indicator, cellulose, and cellulose PEI
  • 60Å silica gel is suitable for all common gel separations including weak to strong polar analytes
  • Silica can also be used for applications requiring charring procedures
  • Aluminum oxide or alumina plates are ideal for separating hormones, alkaloids and antibiotics
  • For partition chromatography, select cellulose plates; and for separation of weak anion exchangers, including peptide and amino acids, use cellulose PEI plates

Certifications

ISO 9001:2000

†With acid-stable fluorescent indicator.

‡PEI (polyethyleneimine) should be stored at 4°C (39°F). If stored at room temperature, PEI plates may turn slightly yellow, which could affect separation. To minimize yellow color, predevelop in distilled water.

Specifications

Backing Material Glass
Certifications/Compliance ISO 9001:2000
Material Silica Gel 60 RP-2 (silanized)
Pore Size 60 Å
Quantity 25/Pk.
Type RP-modified Silica TLC Plate
For Use With (Application) Separation of Extremely Non-Polar and Highly Polar Substances Using Aqueous Solvent Systems
Sorbent Silica Gel 60 RP-2 (silanized)
Thickness (Metric) 250 μm
Dimensions (L x W) 20 x 20 cm
Length (Metric) 20 cm
Width (Metric) 20 cm
Show More Show Less