Learn More
GE Analytical Instruments PRECLEANED GLASS VIALS 30CS
Supplier:  GE Analytical Instruments HMI 7750001
Encompass
Certified Dual Use Conductivity & TOC Vials, 30-mL, 30-case. Pre-cleaned and certified (<10 ppb TOC), no ionic leaching 30-mL specialty coated glass with caps and specialty septa, 30 per case. These vials are suitable for Stage 1 conductivity measurements and simultaneous TOC compliance testing.
Catalog No. NC0943800
Only null left