Learn More
Peak Scientific Instruments Limited Precision Zero Air 7L Produces up to 7L/min of Zero Air
Catalog No. NC1962641
Encompass
Integrates with existing Precision products, with same modular, stackable designHydrocarbon content (as Methane) < 0.05ppm for market-leading purityUp to 7L/min of Zero AirUp to 80psi/5.5bar outlet pressureMinimum maintenance requirements, no expensive catalyst chamber replacementsGreatly reduced risk of contaminants entering system and no risk of running out of gasStylish and contemporary aesthetics and functional designStatus indicative lighting feature (amber warmup, green ready)Compatible with Precision Air Compressor12 month comprehensive on-site warranty
Catalog No. NC1962641 Supplier Peak Scientific Instruments Limited Supplier No. 600008
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Integrates with existing Precision products, with same modular, stackable designHydrocarbon content (as Methane) < 0.05ppm for market-leading purityUp to 7L/min of Zero AirUp to 80psi/5.5bar outlet pressureMinimum maintenance requirements, no expensive catalyst chamber replacementsGreatly reduced risk of contaminants entering system and no risk of running out of gasStylish and contemporary aesthetics and functional designStatus indicative lighting feature (amber warmup, green ready)Compatible with Precision Air Compressor12 month comprehensive on-site warranty