missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Precision Systems NaCl Cryoscope Standard
Description
- For use with milk cryoscope
- Equivalent to 7% sucrose
- -0.422deg.H
- Volume: 250mL
Specifications
Specifications
| Product Type | NaCl Standard |
| For Use With (Equipment) | Milk cryoscope |
| Quantity | 250mL |