missing translation for 'onlineSavingsMsg'
Learn More

Excelta™ Precision Pointed Blade Scissors

Catalog No. 18101002
Change view
Click to view available options
Length (Metric):
118.75 mm
Blade Style:
Angled
Length (Metric) Blade:
31.25 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Blade Style Length (Metric) Blade
18101002 118.75 mm Angled 31.25 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18101002 Supplier Excelta™ Supplier No. 270HT
Only null left

Excelta™ thin pointed blade scissors are made of stainless steel (AISI 440) with carbide blades. While the carbide blades allow for cutting tough material, the slim blades are great for intricate work.

  • Thin pointed blade scissors made of stainless steel (AISI 440) with carbide blades
  • 4 Star Precision
  • While the carbide blades allow for cutting tough material, the slim blades are great for intricate work.

Specifications

Blade Style Angled
Grade 4 Star
Length (English) 4.75 in.
Length (Metric) 118.75 mm
Length (Metric) Blade 31.25 mm
Material Stainless Steel, Carbide
Autoclavable Autoclavable
For Use With (Application) Cutting Tough Material
Length (English) Blade 1.25 in.
Sterility Non-sterile
Tip Style Sharp
Show More Show Less