Learn More
Fisher Container™ Precision Clean™ II Ultra Clear Cleanroom Bags
Excellent clarity, available printed or unprinted
Supplier:  Fisher Container™ 10060A
Encompass
 View more versions of this product
Catalog No. 19033626
Only null left
Description
Description
  • Ideal for packaging semiconductor wafer boxes, medical devices, cleanroom gowns, gloves, wipes or other products requiring a similar level of surface cleanliness
  • Can be sterilized by gamma radiation
  • Clear bags made from ultrapure resin
  • Manufactured and packaged in an ISO Class 4 cleanroom
  • Double-bagged with Certificate of Conformance label
Specifications
Chemical Identifiers
Ultrapure Silicon-free Resin
Specifications
Ultrapure Silicon-free Resin
24 x 18 in. (61 x 45.7cm)
18 in.
45.7 cm
2 mil
24 in.
61 cm
Product Suggestions
Product Suggestions