Learn More
Fisher Container™ Precision Clean™ II Ultra Clear Cleanroom Bags
Excellent clarity, available printed or unprinted
Supplier:  Fisher Container™ FISH0021001
Encompass
 View more versions of this product
Catalog No. 19033616
Only null left
Description
Description
  • Ideal for packaging semiconductor wafer boxes, medical devices, cleanroom gowns, gloves, wipes or other products requiring a similar level of surface cleanliness
  • Can be sterilized by gamma radiation
  • Clear bags made from ultrapure resin
  • Manufactured and packaged in an ISO Class 4 cleanroom
  • Double-bagged with Certificate of Conformance label
Specifications
Chemical Identifiers
Ultrapure Silicon-free Resin
Specifications
Ultrapure Silicon-free Resin
36 x 36 in. (91.4 x 91.4cm)
36 in.
91.4 cm
1.50 mil
36 in.
91.4 cm
Product Suggestions
Product Suggestions