Learn More
Thermo Scientific™ Precision™ Circulating Water Baths
Achieve outstanding temperature uniformity for your samples with perimeter-directed water flow on these circulating water baths. Featuring advanced temperature controller.
Supplier: Thermo Scientific™ TSCIR35
Description
Ensure temperature uniformity and control with theseThermo Scientific™ Precision™ circulating water baths, which is particularly critical when working with enzymes and in serological applications. Available in three different models, these high performance baths feature a microprocessor control and include a removable stainless steel gable cover, diffuser tray, and rubber duck.
- Achieve enhanced temperature uniformity with perimeter-directed water flow
- Easily clean and maintain bath with coil-free internal design
- Optimize scheduling with auto-on and auto-off timers
- Accommodate taller labware with new hinged lid and extended height
- Help prevent bath overheating and damage with low-fluid protection
- Easily operate and monitor with icon-based graphical display
- Protect your work with audible alarms
Chemical Identifiers
| Digital | |
| Digital | |
| Monochrome LCD | |
| 304 Stainless Steel | |
| ±0.1°C | |
| Global Voltage 100/115 V 200/230 V 50/60 Hz |
| UL, CE | |
| Ambient +5° to 100°C | |
| 1 Stainless Steel Gable cover and Rubber Duck | |
| 4 | |
| ±0.05°C |
Specifications
| 9.2 gal. | |
| 35 L | |
| Digital | |
| Monochrome LCD | |
| 1 Stainless Steel Gable cover and Rubber Duck | |
| Precision CIR 35 | |
| ±0.1°C | |
| 12 x 27.3 x 7.6 in. (305 x 692 x 192 mm) | |
| Global Voltage 100/115 V 200/230 V 50/60 Hz |
| Digital | |
| UL, CE | |
| Ambient +5° to 100°C | |
| 1500 w | |
| 304 Stainless Steel | |
| 4 | |
| ±0.05°C | |
| 15.5 x 36.9 x 9.8 in. (394 x 938 x 249 mm) | |
| 11.2-13.1 / 5.4-6.3 (120V/230V) |