missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Precision™ Circulating Water Baths

Catalog No. TSCIR19
Change view
Click to view available options
Capacity (Metric):
19 L
35 L
89 L
Model:
Precision CIR 19
Precision CIR 35
Precision CIR 89
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Model
TSCIR19 19 L Precision CIR 19
TSCIR35 35 L Precision CIR 35
TSCIR89 89 L Precision CIR 89
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. TSCIR19 Supplier Thermo Scientific™ Supplier No. TSCIR19
  • ONLINE EXCLUSIVE PRICE
    $5,552.99
    $4,523.05 / Each

    Save $1,029.94

    19.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Achieve outstanding temperature uniformity for your samples with perimeter-directed water flow on these circulating water baths. Featuring advanced temperature controller.

Ensure temperature uniformity and control with theseThermo Scientific™ Precision™ circulating water baths, which is particularly critical when working with enzymes and in serological applications. Available in three different models, these high performance baths feature a microprocessor control and include a removable stainless steel gable cover, diffuser tray, and rubber duck.

  • Achieve enhanced temperature uniformity with perimeter-directed water flow
  • Easily clean and maintain bath with coil-free internal design
  • Optimize scheduling with auto-on and auto-off timers
  • Accommodate taller labware with new hinged lid and extended height
  • Help prevent bath overheating and damage with low-fluid protection
  • Easily operate and monitor with icon-based graphical display
  • Protect your work with audible alarms

Specifications

Capacity (English) 5.0 gal.
Controller Type Digital
Capacity (Metric) 19 L
Certifications/Compliance UL, CE
Communications Digital
Temperature Range Ambient +5° to 100°C
Display Monochrome LCD
Heating Capacity 1200 w
Includes 1 Stainless Steel Gable cover and Rubber Duck
Material 304 Stainless Steel
Model Precision CIR 19
Temperature Presets 4
Temperature Stability ±0.1°C
Temperature Uniformity ±0.05°C
Dimensions (D x W x H) Interior 12 x 15.3 x 7.6 in. (305 x 387 x 192 mm)
Dimensions (L x W x H) 15.5 x 24.9 x 9.8 in. (394 x 632 x 249 mm)
Electrical Requirements Global Voltage 100/115 V 200/230 V 50/60 Hz
Amperage 9.0-10.5 / 4.3-5.0 (120V/230V)
Show More Show Less