missing translation for 'onlineSavingsMsg'
Learn More

Precipitated Sulfur, 99.5-100.5%, Spectrum™ Chemical

Catalog No. p-7158087
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605783 500 g Poly Bottle
18605784 2.5 kg Poly Pail
18605785 12 kg Poly Pail
18605786 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18605783 Supplier Spectrum Chemical Mfg Cor Supplier No. S1384500GM
Only null left

S1384, 7704-34-9, S

Spectrum™ Chemical Precipitated Sulfur is an abundant, non-metal, multivalent chemical element. In cosmetics and personal care products Sulfur is used to enhance the appearance and feel of hair, by increasing hair body, suppleness, or sheen, or by improving the texture of hair that has been damaged physically or by chemical treatment and to enhance the appearance of dry or damaged skin by reducing flaking and restoring suppleness.

Chemical Identifiers

CAS 7704-34-9
Molecular Formula S
Molecular Weight (g/mol) 32.06
MDL Number MFCD00085316
InChI Key NINIDFKCEFEMDL-UHFFFAOYSA-N
IUPAC Name sulfur
SMILES [S]

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 99.5 to 100.5%
Grade Reagent
Packaging Poly Bottle