missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ambion™ Pre-miR™ miRNA Precursor
Description
miRNA Precursor Molecules are small, chemically modified double-stranded RNA molecules designed to mimic endogenous mature miRNAs.
Specifications
Specifications
| Concentration | 5X |
| Content And Storage | Pre-miR™ miRNA is provided dried down in a single tube, along with Nuclease-free Water for resuspension, and should be stored at –20°C. It is guaranteed for 1 year when stored dried at –20°C. |
| Description | Pre-miR™ miRNA Precursor Molecule, 5nmol, Liquid, Protect Against Physical Damage, Isolate From Incompatible Substances, Store in Original Container |
| Final Product Type | Pre-miR™ miRNA Precursor Molecule |
| Packaging Type | 1 Each |
| Product Line | Ambion, Pre-miR |
| Product Type | Pre-miR™ miRNA Precursor Molecule |
| Quantity | 5 nmol |
| Sample Size | 5nmol |
| Form | Liquid |
For Research Use Only. Not for use in diagnostic procedures.