missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Pre-miR™ hsa-miR-1 miRNA Precursor Positive Control

Catalog No. AM17150
Encompass
Change view
Click to view available options
Quantity:
5 nmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM17150 5 nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM17150 Supplier Invitrogen™ Supplier No. AM17150

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Pre-miR™ hsa-miR-1 miRNA Precursor is intended for use as a positive control in experiments utilizing Pre-miR miRNA Precursors.

The Pre-miR™ hsa-miR-1 miRNA Precursor is intended for use as a positive control in experiments utilizing Pre-miR miRNA Precursors. This precursor was chosen as a positive control because, upon delivery into cells, it effectively down regulates the expression of twinfilin-1, also known as PTK9, at the mRNA level (60-95% reduction. Effective delivery and activity of the Pre-miR hsa-miR-1 miRNA Precursor can be detected by real-time PCR using a TaqMan™ Gene Expression Assay to PTK9 (Assay IDs: Hs00702289_s1 [human], Mm01598980_g1 [mouse]).

About Pre-miR miRNA Precursors
Pre-miR™ miRNA Precursor Molecules are small, chemically modified double-stranded RNA molecules designed to mimic endogenous mature miRNAs. These ready-to-use miRNA mimics, which are similar to, but not identical to siRNAs, can be introduced into cells using transfection or electroporation parameters similar to those used for siRNAs. They enable detailed study of miRNA biological effects via gain-of-function experiments (Figure 1). Positive and negative control Pre-miR miRNA Precursors (listed above) or the Pre-miR™ Precursor Starter Kit can be used to optimize transfection of Pre-miR miRNA Precursors.

Pre-miR miRNA Precursor Molecules have been carefully designed and modified to ensure that the correct strand, representing the desired mature miRNA, is taken up into the RNA-induced silencing-like complex responsible for miRNA activity (Figure 2). In contrast to miRNA expression vectors, these synthetic molecules can be used in dose response studies due to their introduction directly into the cell by transfection or electroporation. Thanks to their small size, they are easier to transfect than vectors, and can be delivered using conditions identical to those used for siRNAs.

Specifications

Content And Storage Pre-miR™ miRNA is provided dried down in a single tube, along with Nuclease-free Water for resuspension, and should be stored at -20°C. Guaranteed for 1 year when stored dried at -20°C.
Control Type Positive Control
RNAi Type miRNA Mimic
Species Human, Mouse, Rat
For Use With (Application) miRNA & Non-Coding RNA Analysis
Includes Pre-miR™ miRNA is provided dried down in a single tube, along with nuclease-free Water for resuspension
Label or Dye Unconjugated
Product Line Ambion, Pre-miR
Product Type miRNA Mimic
Purity HPLC
Quantity 5 nmol
Shipping Condition Room Temperature
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.