missing translation for 'onlineSavingsMsg'
Learn More

StatLab™ Pre-cut Cotton Gauze

Pre-cut cotton gauze in 4 x 4 in. squares.

Supplier:  StatLab™ 7634

 View more versions of this product

Catalog No. STL7634


Only null left
Explore available promotions

Description

Description

  • For general laboratory use or for coverslipping
  • Non-sterile
Specifications

Specifications

Polyester/Viscose Blend
Non-sterile
10.2 cm
4 in.
Medium
Not Autoclavable
Gauze Pad
4 in.
10.2 cm
200 Bandages/Pk.
Yes
4-ply
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions