Learn More
Thermo Scientific™ Pravastatin sodium salt hydrate
Chemical Identifiers
| CAS | 2244386-63-6 |
|---|---|
| Molecular Formula | C23H35O7Na·xH2O |
| Molecular Weight (g/mol) | 446.51 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC464880010
|
Thermo Scientific™
464880010 |
1 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC464882500
|
Thermo Scientific™
464882500 |
250 mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 2244386-63-6 | |
| C23H35O7Na·xH2O | |
| 446.51 |
Specifications
| 2244386-63-6 | |
| 1 g | |
| 446.51 | |
| Pravastatin sodium salt hydrate |
| C23H35O7Na·xH2O | |
| Solubility aq. soln.: soluble. | |
| 446.51 |
Safety and Handling
GHS H Statement Suspected of damaging fertility or the unborn child.
May cause harm to breast-fed children.
GHS P Statement Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
Avoid contact during pregnancy and while nursing
GHS Signal Word: Warning
RUO â Research Use Only