missing translation for 'onlineSavingsMsg'
Learn More

Prasugrel 98.0+%, TCI America™

Catalog No. P2040200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2040200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P2040200MG Supplier TCI America Supplier No. P2040200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 150322-43-3
Molecular Formula C20H20FNO3S
Molecular Weight (g/mol) 373.442
MDL Number MFCD09954140
InChI Key DTGLZDAWLRGWQN-UHFFFAOYSA-N
Synonym prasugrel, effient, efient, prasugrel effient, prasita, 5-2-cyclopropyl-1-2-fluorophenyl-2-oxoethyl-4,5,6,7-tetrahydrothieno 3,2-c pyridin-2-yl acetate, 2-2-acetyloxy-6,7-dihydrothieno 3,2-c pyridin-5 4h-yl-1-cyclopropyl-2-2-fluorophenyl ethanone, effient tn, ethanone, 2-2-acetyloxy-6,7-dihydrothieno 3,2-c pyridin-5 4h-yl-1-cyclopropyl-2-2-fluorophenyl
PubChem CID 6918456
ChEBI CHEBI:87723
IUPAC Name [5-[2-cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl]-6,7-dihydro-4H-thieno[3,2-c]pyridin-2-yl] acetate
SMILES CC(=O)OC1=CC2=C(S1)CCN(C2)C(C3=CC=CC=C3F)C(=O)C4CC4

Specifications

Melting Point 122°C
Quantity 200 mg
Formula Weight 373.44
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Prasugrel
TSCA No
Recommended Storage Refrigerator