missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium rod, 6.35mm (0.25in) dia, 98.5% (metals basis excluding Ta)

Catalog No. p-4782091
Change view
Click to view available options
Quantity:
10 mm
25 mm
50 mm
100 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40295HF 10 mm
AA40295DH 25 mm
AA40295DJ 50 mm
AA40295DM 100 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA40295HF Supplier Thermo Scientific Chemicals Supplier No. 040295HF
Only null left

Form: Approx. 0.22g/mm, Cast rod, diameter is approximate | CAS: 7440-10-0 | Pr | 140.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-10-0
Molecular Formula Pr
Molecular Weight (g/mol) 140.91
MDL Number MFCD00011174
InChI Key PUDIUYLPXJFUGB-UHFFFAOYSA-N
Synonym unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg
PubChem CID 23942
ChEBI CHEBI:49828
IUPAC Name praseodymium
SMILES [Pr]

Specifications

Color Silver
Physical Form Cast Rod
Quantity 10 mm
Assay Percent Range 99.10%
Odor Odorless
Appearance Silver colored rod
Chemical Name or Material Praseodymium
EINECSNumber 231-120-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only