missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pramiracetam Hydrate 98.0+%, TCI America™
$70.84 - $342.98
Chemical Identifiers
| CAS | 68497-62-1 |
|---|---|
| Molecular Formula | C14H27N3O2 |
| Molecular Weight (g/mol) | 269.389 |
| MDL Number | MFCD00867219 |
| InChI Key | ZULJGOSFKWFVRX-UHFFFAOYSA-N |
| PubChem CID | 51712 |
| IUPAC Name | N-[2-[di(propan-2-yl)amino]ethyl]-2-(2-oxopyrrolidin-1-yl)acetamide |
| SMILES | CC(C)N(CCNC(=O)CN1CCCC1=O)C(C)C |
Chemical Identifiers
| 68497-62-1 | |
| 269.389 | |
| ZULJGOSFKWFVRX-UHFFFAOYSA-N | |
| N-[2-[di(propan-2-yl)amino]ethyl]-2-(2-oxopyrrolidin-1-yl)acetamide |
| C14H27N3O2 | |
| MFCD00867219 | |
| 51712 | |
| CC(C)N(CCNC(=O)CN1CCCC1=O)C(C)C |
Specifications
| 68497-62-1 | |
| White-Yellow | |
| C14H27N3O2 | |
| 1 g | |
| CC(C)N(CCNC(=O)CN1CCCC1=O)C(C)C | |
| 269.389 | |
| 269.39 | |
| Crystalline Powder |
| 47°C | |
| 164°C | |
| MFCD00867219 | |
| ZULJGOSFKWFVRX-UHFFFAOYSA-N | |
| N-[2-[di(propan-2-yl)amino]ethyl]-2-(2-oxopyrrolidin-1-yl)acetamide | |
| 51712 | |
| ≥98.0% (HPLC,T) | |
| Pramiracetam Hydrate |
Safety and Handling
RTECSNumber : UV6264081
TSCA : No