missing translation for 'onlineSavingsMsg'
Learn More

Pramiracetam Hydrate 98.0+%, TCI America™

Catalog No. P20611G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P20611G 1 g
P20615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P20611G Supplier TCI America Supplier No. P20611G
Only null left

Chemical Identifiers

CAS 68497-62-1
Molecular Formula C14H27N3O2
Molecular Weight (g/mol) 269.389
MDL Number MFCD00867219
InChI Key ZULJGOSFKWFVRX-UHFFFAOYSA-N
PubChem CID 51712
IUPAC Name N-[2-[di(propan-2-yl)amino]ethyl]-2-(2-oxopyrrolidin-1-yl)acetamide
SMILES CC(C)N(CCNC(=O)CN1CCCC1=O)C(C)C

Specifications

Melting Point 47°C
Color White-Yellow
Boiling Point 164°C
Quantity 1 g
Formula Weight 269.39
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pramiracetam Hydrate
RTECSNumber UV6264081
TSCA No