missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pramipexole Dihydrochloride Monohydrate 98.0+%, TCI America™
$52.19 - $283.33
Chemical Identifiers
| CAS | 191217-81-9 |
|---|---|
| Molecular Formula | C10H21Cl2N3OS |
| Molecular Weight (g/mol) | 302.26 |
| MDL Number | MFCD00876894,MFCD02183927 |
| InChI Key | APVQOOKHDZVJEX-QTPLPEIMSA-N |
| Synonym | (S)-2-Amino-4,5,6,7-tetrahydro-6-(propylamino)benzothiazole Dihydrochloride |
| PubChem CID | 166589 |
| ChEBI | CHEBI:51147 |
| IUPAC Name | dihydrogen (6S)-N6-propyl-4,5,6,7-tetrahydro-1,3-benzothiazole-2,6-diamine hydrate dichloride |
| SMILES | [H+].[H+].O.[Cl-].[Cl-].CCCN[C@H]1CCC2=C(C1)SC(N)=N2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P2073100MG
|
TCI America
P2073100MG |
100 mg |
Each for $52.19
|
|
|||||
|
P20731G
|
TCI America
P20731G |
1 g |
Each for $283.33
|
|
|||||
Chemical Identifiers
| 191217-81-9 | |
| 302.26 | |
| APVQOOKHDZVJEX-QTPLPEIMSA-N | |
| 166589 | |
| dihydrogen (6S)-N6-propyl-4,5,6,7-tetrahydro-1,3-benzothiazole-2,6-diamine hydrate dichloride |
| C10H21Cl2N3OS | |
| MFCD00876894,MFCD02183927 | |
| (S)-2-Amino-4,5,6,7-tetrahydro-6-(propylamino)benzothiazole Dihydrochloride | |
| CHEBI:51147 | |
| [H+].[H+].O.[Cl-].[Cl-].CCCN[C@H]1CCC2=C(C1)SC(N)=N2 |
Specifications
| 191217-81-9 | |
| White-Yellow | |
| MFCD00876894,MFCD02183927 | |
| (S)-2-Amino-4,5,6,7-tetrahydro-6-(propylamino)benzothiazole Dihydrochloride | |
| [H+].[H+].O.[Cl-].[Cl-].CCCN[C@H]1CCC2=C(C1)SC(N)=N2 | |
| 302.26 | |
| CHEBI:51147 | |
| ≥98.0% (HPLC,N) | |
| Pramipexole Dihydrochloride Monohydrate |
| 290°C | |
| C10H21Cl2N3OS | |
| 100 mg | |
| APVQOOKHDZVJEX-QTPLPEIMSA-N | |
| dihydrogen (6S)-N6-propyl-4,5,6,7-tetrahydro-1,3-benzothiazole-2,6-diamine hydrate dichloride | |
| 166589 | |
| 284.24 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : DL3375000
TSCA : No