Learn More
Eisco™ Practice Suture Kit - 5 Pack
Catalog No. S02942
Change view
Click to view available options
Product Type: Complete Practice Kit
Catalog No.
S02942 Product Type: Complete Practice Kit
1 options
Catalog No. S02942 Supplier Eisco™ Supplier No. BI0201903PK5
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Premium, life-like plastic suture pad with common cuts and lacerations - needle holder, forceps, scissors, tubing, needles and sutures. Designed by a physician, the Practice Suture Kit includes everything needed to practice suturing skills, including a variety of wound types and needle sizes.
  • The Eisco Labs 5 pack of Suture Practice Kits includes everything you need to practice suturing skills, including various suture types and needle sizes
  • The kit features several different practice cuts, and has been designed for use by students in medical, nursing and veterinary schools, as well as surgical, emergency and anesthesia residents
  • The kit is compact and can be used for practicing suturing techniques at home or demonstrating techniques in the classroom or laboratory
  • Our innovative plastic suture pad is durable and does not tear, allowing each cut to be sutured multiple times
  • All the kit components can be conveniently stored in the sturdy box
  • Suture pad measures 6.5 in. wide, 5.75 in. long and 0.5 in. thick. Included in each kit:
  • (1) Suture pad with wounds
  • (1) Needle holder
  • (1) Toothed forceps for handling tissue
  • (1) Fine suturing scissors
  • (5) Braided Silk Suture Varieties - 0/0, 2/0, 3/0, 4/0, and 5/0
  • (4) Half circle needles, 19 mm
  • (1) Half circle needle, 37 mm
  • (2) Tubing pieces to practice securing chest tubes, drains, central lines, arterial and umbilical catheters to the skin
All contents in the kit are for practice and educational use only. They should never be used on a living specimen of any kind.
Specifications
Product Type Complete Suture Practice Kit
For Use With (Application) Suturing Skills