missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ PR+ Field Installable Cutter CU4
Description
- Package includes Replacement Printer Cutter only (printer sold separately)
- 4-inch blade designed for use with Bradyprinter™ PR300 Plus and PR600 Plus Printer Series (mfr model numbers BP-PR300+ and BP-PR600+)
- Sharp cutter blades cut easily through label materials
- No need to carry around extra scissors when a cutter is build into the printer
Specifications
Specifications
| Product Type | PR+ Field Installable Cutter CU4 |
| Color | Black |
| For Use With (Equipment) | BradyPrinter PR Plus Series Label Printer |
| Sterility | Non-sterile |