missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Protein Precipitation Solution (PPS) for Soil Kit
Description
- Applications: PPS is component of FastDNA™ SPIN Kit for Soil.
- Usage Statement:
- Research Use Only (RUO).
- Ready for CE IVD labeling of clinical applications.
- All Fast DNA Kit reagents are stable at room temperature(15°C to 30°C).
- Shelf life can be extended by storing at 4°C.
Specifications
Specifications
| For Use With (Application) | Protein Precipitation Solution |
| For Use With (Equipment) | All FastPrep Adapters |
| Packaging Quantity | 25 mL |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.