missing translation for 'onlineSavingsMsg'
Learn More

Cupric Chloride, Dihydrate, Crystal, ACS, 99%, Spectrum™ Chemical

Catalog No. 18609996
Change view
Click to view available options
Quantity:
5 g
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609996 50 kg Amber Glass Bottle
18609990 5 g Amber Glass Bottle
18609991 25 g Amber Glass Bottle
18609992 125 g Amber Glass Bottle
18609993 500 g Amber Glass Bottle
18609994 2.5 kg Amber Glass Bottle
18609995 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18609996 Supplier Spectrum Chemical Mfg Cor Supplier No. C138550KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10125-13-0 | 170.48 g/mol

Chemical Identifiers

CAS 10125-13-0

Specifications

CAS Max % 1
Physical Form Crystals or Needles
Quantity 50 kg
Molecular Weight (g/mol) 170.48
Percent Purity 99%
Grade ACS/Reagent
Packaging Amber Glass Bottle
Chemical Name or Material Cupric Chloride, Dihydrate, Crystal

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.