Learn More
MilliporeSigma™ PP2A Immunoprecipitation Phosphatase Assay Kit
Catalog No. 17313
Change view
Click to view available options
Quantity:
25 Assays
Catalog No. Quantity
17313 25 Assays
1 options
Catalog No. 17313 Supplier MilliporeSigma™ Supplier No. 17313
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
  • Malachite Green Solution A
  • Malachite Green Additive
  • Phosphate Standard
  • Threonine Phosphopeptide (K-R-pT-I-R-R)
  • Anti-PP2A, C subunit, clone 1D6
  • pNPP Ser/Thr Assay Buffer
  • Protein A Agarose
  • 96-Well Microtiter Plate
Utilizes Malachite Green and contains components that have been matched to optimize sensitivity and range of detection
  • Can be used to detect purified PP2A or PP2A from a cellular lysate
  • It utilizes the Malachite Green and contains components that have been matched to optimize sensitivity and range of detection
  • Application: Phosphatase Assay
Specifications
For Use With (Application) Phosphatase Assay
Quantity 25 Assays