Learn More
Bel-Art™ Poxygrid™ Biohazard Bag Holders
Sturdy wire holder keeps bags open while a separate hooked ring that rests over the top keeps them securely in place
Supplier:  Bel-Art™ H131880000
 View more versions of this product
Catalog No. 0181510B
Get More Bel-Art Lab Products with Purchase
View all promotions
Only null left
Description
Description
Sturdy wire holder keeps bags open while a separate hooked ring that rests over the top keeps them securely in place
  • Bright biohazard orange colored epoxy-coated steel wire
  • Chemical and corrosion-resistant; unaffected by organic solvents, salts, weak acids and weak alkalis
  • Will withstand a temperature range of -223°C (-369°F) to 121°C (250°F)
  • Autoclavable
Specifications
Chemical Identifiers
Circular epoxy coated wire frame with easy-to-use hooked ring over a top
Orange
To keep biohazard bags open and ready for use
-369°F to +250°F
Biohazard Bag Holder
Autoclavable
Reusable
Epoxy-coated Wire
-223°C to +121°C
Specifications
Circular epoxy coated wire frame with easy-to-use hooked ring over a top
Orange
To keep biohazard bags open and ready for use
Epoxy-coated Wire
-223°C to +121°C
23.5 in.
14.6 in.
Biohazard Bag Holder
Autoclavable
Reusable
01-814-2, 01-814C, 01-815B, 01-826-6, 01-826B, 01-827C, 01-830D, 01-831A, 01-828-033, 01-832A 30 H x 24 in. W biohazard bags
-369°F to +250°F
14.6 in.
60 cm
37 cm
37 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Accessories