OptiChem Heating Mantle Controller, Double Circuit, 7A, 840 Watts, (per Circuit), 120VAC. Our OptiChem line of heating mantle controllers feature solid-state proportional voltage power control in a compact benchtop design. Controllers provide accurate manual control of heating mantles, tapes and all other resistive loads with an adjustable output range from 5 to 100% of the rated voltage. Controller is 8 W x 6 D x 3.7 H. All units are supplied with a 6ft separable power cord, a grounded, 3-wire receptacle and lighted power switch. 3-Year warranty.OptiChem is a Registered Trademark of Chemglass, Inc.