missing translation for 'onlineSavingsMsg'
Learn More

SW Sustainability Solutions PowerForm PF-95GW 5.9 mil Sustainable Skin-Improving Nitrile Exam Gloves
GREENER_CHOICE

Catalog No. p-7237835
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17441029 X-Small
17441030 Small
17441031 Medium
17441032 Large
17441033 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17441029 Supplier SW Sustainability Solutions Supplier No. N189471
Only null left

EnerGel infused gloves are designed to hydrate skin and protect against long-term glove usage.

  • EnerGel Hand Conditioning Technology hydrates and soothes skin, proven to prevent many skin ailments
  • EcoTek Sustainable Technology
  • Breach Alert Visual Detection Technology provides durable 2-ply construction and contrasting inner color
  • Proven resistance to fentanyl and gastric acid for over 4 hours to replicate hazardous settings and overdose conditions
  • Fully textured for secure grip
  • Silicone-free for critical environments
  • 1.5 AQL exceeds laboratory standards

Specifications

Certifications/Compliance ASTM D6319-19, ASTM D6978-05 (2019), ASTM F1671-13, ASTM F739-12
Certifications/Compliance (Europe) EN ISO 21420:2020, EN 455-2000-2015, Reach EU 1907/2006, EU 2016/425
Chemotherapeutic Rated Yes
Chlorinated Yes
Accelerator Free No
Applications Healthcare, Laboratory, Industrial
AQL 1.5
Color Green, White
Disposable Single-use
Exterior Finish Fully Textured
Finger Thickness (English) 5.9 mil
Finger Thickness (Metric) 0.15 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 4.9 mil
Palm Thickness (Metric) 0.13 mm
Powdered Powder Free
Product Type Nitrile Examination Glove
Size X-Small
Sterility Non-sterile
Thickness (English) 5.9 mil
Thickness (Metric) 0.15 mm
FDA 510 (k) Yes
Glove Cuff Style Beaded
Model N189471
Series PF-95GW
Tensile Strength 18 MPa
Length (English) 9.5 in.
Length (Metric) 24.2 cm
Show More Show Less

*EcoTek gloves tested to ASTM D5526-12 Standard Test Method for Determining Anaerobic Biodegradation of Plastic Materials Under Accelerated Landfill Conditions. Results are 92.6% biodegradation in 60% solids landfill at 945 days. Future results cannot be predicted/extrapolated.