missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PowerEase™ Touch HV Power Supply and ZOOM™ IPGRunner Welcome Pack

Catalog No. PSHV00Z
Change view
Click to view available options
Quantity:
1 each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PSHV00Z 1 each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PSHV00Z Supplier Invitrogen™ Supplier No. PSHV00Z
Only null left

The PowerEase Touch HV Power Supply and ZOOM IPGRunner Welcome Pack contains all of the reagents and equipment needed for isoelectric focusing of protein samples when purchased with Zoom IPG strips and ampholytes.

The PowerEase Touch HV Power Supply and ZOOM IPGRunner Welcome Pack contains all of the reagents and equipment needed for isoelectric focusing of protein samples when purchased with Zoom IPG strips and ampholytes. Zoom IPG strips and ampholytes are sold separately in different pH ranges for proper separation of the proteins being studied.

The PowerEase Touch HV Power Supply and ZOOM IPGRunner Welcome Pack contains:
• Power Ease Touch HV Power Supply (Cat. No. varies with region)
• ZOOM IPGRunner Mini-cell (Cat. No. ZM0001)
• ZOOM IPGRunner Cassette (Cat. No. ZM0003)
• ZOOM Urea (Cat. No. ZU10001)
• ZOOM Thiourea (Cat. No. ZT10002)
• ZOOM CHAPS (Cat. No. ZC10003)
• ZOOM Equilibration Trays (ZM0007)

PowerEase Touch HV Power Supply
The Invitrogen PowerEase Touch HV Power Supply is a high-voltage power supply that is operated using a touch-screen user interface. The 4.3-inch backlit LCD touchscreen and intuitive user interface design make setting up custom protocols or selecting one of the several pre-programmed gel electrophoresis and transfer methods a breeze. This power supply is ideal for high-voltage applications like 2D electrophoresis or isoelectric focusing. It also supports high-throughput DNA/RNA electrophoresis, SDS-PAGE, native PAGE, and western blotting.

The PowerEase Touch HV Power Supply offers four sets of output jacks that can be used simultaneously and four operation modes: constant voltage, voltage ramp, constant current, and constant power for flexibility of use and efficiency.

ZOOM IPGRunner system
The ZOOM IPGRunner system is an oil-free, 2D electrophoresis system in a mini-gel format. The first dimension separation by isoelectric focusing (IEF) is typically complete in less than three hours, and the system allows for processing of up to 12 samples at once for high-throughput usage using ZOOM IPG Strips tailored to the pH range of the protein(s) of interest. When combined with Zoom IPG Strips this welcome pack contains all of the equipment and reagents required for performing first-dimension separation for 2D electrophoresis or isoelectric focusing within the IPG strip format.

Second dimension analysis
For second dimension separation we recommend the NuPAGE Bis-Tris XCell SureLock Mini-Cell Welcome Pack, 4–12%, IPG-well (Cat. No. NP0330Z), which contains everything needed for your second-dimension separation by size including the XCell SureLock Mini-Cell tank.

Specifications

Content And Storage • PowerEase™ Touch HV Power Supply
• ZOOM IPGRunner Buffer Core and Lid
• Gel Tension Wedge
• Buffer Dam
• Mini-Cell chamber

For Use With (Application) Gel Electrophoresis, Isoelectric Focusing (IEF or IPG)
For Use With (Equipment) XCell SureLock Mini, ZOOM IPGRunner Mini
Gel Compatibility ZOOM™ IPG Strips
Gel Size 7 cm Strips
Product Line IPGRunner, ZOOM
Quantity 1 each
Do you offer a power supply that is comparable to the Bio-Rad PowerPac HV Power Supply?

Yes, we recommend using the PowerEase Touch HV Power Supply.

Do you have an alternative for the discontinued ZOOM Dual Power Supply?

We offer the PowerEase Touch HV Power Supply as an alternative for the discontinued ZOOM Dual Power Supply.

Do I need to use PowerEase Touch HV Power Supply Adapters with the PowerEase Touch HV Power Supply even if I have Invitrogen gel tanks?

The PowerEase Touch HV Power Supply Adapters are designed to facilitate the connection of some electrophoresis tanks to the PowerEase Touch HV Power Supply. The adapters are designed to remain on the electrophoresis tank electrical leads when the lid is removed from the power supply. The supported electrophoresis tanks include:

- ZOOM IPGRunner Mini-cell
- XCell SureLock Mini-Cell
- Mini Gel Tank
- XCell4 SureLock Midi-Cell
Note: The PowerEase Touch HV Power Supply Adapters are not needed for use of the SureLock Tandem Midi Gel Tank with the PowerEase Touch HV Power Supply.

Why does the PowerEase Touch HV Power Supply come with power supply adapters?

For safety purposes, the output jacks of the PowerEase Touch HV Power Supply have been recessed compared to other PowerEase Touch power supplies. This requires a firm elongation of the leads on the cables of the tank and this is achieved using the power supply adapters that are provided. The adapters are designed to remain on the electrophoresis tank electrical leads when the lid is removed from the power supply.

How many gels can I simultaneously transfer using the PowerEase Touch HV Power Supply?

The PowerEase Touch HV Power Supply can be used for simultaneous transfer of 2 mini gels. We do not recommend using it for transfer of midi gels.

How many ZOOM IPG Strips can I simultaneously run using the PowerEase Touch HV Power Supply?

The PowerEase Touch HV Power Supply can be used for simultaneous running of 48 ZOOM IPG Strips.

How many gels can I simultaneously run using the PowerEase Touch HV Power Supply?

The PowerEase Touch HV Power Supply can be used for simultaneous running of 8 IEF gels, 4 mini gels, or 2 midi gels for SDS-PAGE.

Which applications can be run using the PowerEase Touch HV Power Supply?

The PowerEase Touch HV Power Supply is ideal for high voltage separation such as isoelectric focusing (IEF gels and IPG Strips) and 2-D electrophoresis. In addition, it also supports DNA/RNA electrophoresis.

Can the PowerEase Touch 600W Power Supply be used to run ZOOM IPG strips?

No, ZOOM IPG strips require a voltage ramp up to 2000 V to be run. We recommend using the PowerEase Touch HV Power Supply to run ZOOM IPG Strips.

Can I use the PowerEase Touch 120W or PowerEase Touch 350W Power Supply to run isoelectric focusing (IEF) gels?

The voltage required for running of IEF gels cannot be reached with the PowerEase Touch 120W or the PowerEase Touch 350W Power Supply. We recommend using the PowerEase Touch 600W Power Supply or PowerEase Touch HV Power Supply for running IEF gels.


For Research Use Only. Not for use in diagnostic procedures.