missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PowerEase™ Touch 350W Power Supply and XCell SureLock™ Mini-Cell and XCell II™ Blot Module Welcome Pack DFS Item

The PowerEase Touch 350W Power Supply and XCell SureLock Mini-Cell and XCell II Blot Module Welcome Pack contains: PowerEase Touch 350W Power Supply and XCell SureLock Mini-Cell and XCell II Blot Module

Supplier:  Invitrogen™ PSC350SB

Catalog No. PSC350SB

Only null left
Explore available promotions

Description

Description

The PowerEase Touch 350W Power Supply and XCell SureLock Mini-Cell and XCell II Blot Module Welcome Pack contains:

  • PowerEase Touch 350W Power Supply
  • XCell SureLock Mini-Cell and XCell II Blot Module

About the PowerEase Touch 350W Power Supply

The Invitrogen PowerEase Touch 350W Power Supply is a high-throughput power supply with a 4.3-inch backlit LCD touchscreen. The touchscreen and user interface design make setting up custom protocols or selecting one of the several pre-programmed gel electrophoresis and transfer methods a breeze. This power supply is ideal for high current applications like high throughput DNA/RNA electrophoresis, SDS-PAGE, native PAGE, and western blotting. The PowerEase Touch 350W Power Supply offers four sets of output jacks that can be used simultaneously and three modes: constant voltage, constant current, and constant power for flexibility of use and efficiency.

About the XCell SureLock Mini-Cell and XCell II Blot Module

The XCell SureLock Mini-Cell and XCell II Blot Module set is an electrophoresis and blotting system for mini gels. It includes the XCell SureLock Mini-Cell for running Invitrogen mini gels and the XCell II Blot Module for blotting up to two mini gels at a time. The XCell SureLock Mini-Cell holds gels firmly with a simple gel tension wedge. When the lever on the gel tension wedge is pushed forward into the locked position, an even, horizontal force is generated. This seals the gel/buffer core assembly firmly into position in the lower buffer chamber. The positive locking action of the gel tension wedge ensures a trouble-free, leak-free gel run every time.

The XCell II Blot Module allows you to easily transfer proteins or nucleic acids from mini gels to membranes. It fits into the XCell SureLock and XCell II Mini-Cells in place of the gel/buffer core assembly. It requires less than 200 mL of transfer buffer for western, southern, and northern transfers. Tough platinized titanium and stainless steel electrodes create a uniform electrical field without clamps or hinged gel holders. Maximum blot size is 9 cm x 9 cm.

Specifications

Specifications

Room Temperature
Mini
PowerEase
4.3 inch backlit LCD
1 pack
Yes
30.3 inch x 23 inch x 10.2 inch
300 VDC
High throughput DNA/RNA electrophoresis, SDS-PAGE, native PAGE and western blotting
3 A
Up to 999 min./step
4 (4 positive voltage and 4 negative voltage)
Room Temperature
One year
2.1 kg
350 watts
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions